Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furan-3-boronic acid, 97%, Acros Organics

Click to view available options
Quantity:
100mg
500mg
2.5g
Packaging:
Glass bottle
55552-70-0
C4H5BO3
111.89
MFCD01319007
CYEFKCRAAGLNHW-UHFFFAOYSA-N
furan-3-boronic acid,3-furanboronic acid,3-furylboronic acid,furan-3-yl boronic acid,3-furanylboronic acid,furan-3-ylboranediol,3-furan boronic acid,furan-3-yl-3-boronic acid,furan-3-yl-boronic acid,boronic acid, 3-furanyl
2734358
furan-3-ylboronic acid
B(C1=COC=C1)(O)O
55552-70-0
C4H5BO3
111.89
MFCD01319007
CYEFKCRAAGLNHW-UHFFFAOYSA-N
furan-3-boronic acid,3-furanboronic acid,3-furylboronic acid,furan-3-yl boronic acid,3-furanylboronic acid,furan-3-ylboranediol,3-furan boronic acid,furan-3-yl-3-boronic acid,furan-3-yl-boronic acid,boronic acid, 3-furanyl
2734358
furan-3-ylboronic acid
B(C1=COC=C1)(O)O
Melting Point 126°C to 128°C
Packaging Glass bottle
Quantity 100mg
Infrared Spectrum Authentic
Color Yellow
Assay Percent Range 96% min. (HPLC)
Formula Weight 111.89
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Furan-3-boronic acid

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning