Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Furan, 99+%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -85.6°C |
Color | Colorless to Yellow |
Density | 0.9360g/mL |
Boiling Point | 31.3°C |
Flash Point | −35°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.05% max. (Furfural) (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4197 to 1.4217 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. Harmful if inhaled. May cause cancer. Harmful to aquatic life with long |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the |
EINECSNumber | 203-727-3 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction