Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furazolidone, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

F2272, 67-45-8, C8H7N3O5

Spectrum™ Furazolidone, USP is used to treat bacterial and protozoal infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.25%
Loss on Drying 1%
Packaging Fiber Drum
Quantity 25 kg
CAS 67-45-8
CAS Max % 100%
Molecular Formula C8H7N3O5
InChI Key PLHJDBGFXBMTGZ-WEVVVXLNSA-N
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CCOC1=O
IUPAC Name 3-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 225.16
Percent Purity 97 to 103%
Grade USP
Show More Show Less