Alfa Aesar™ Furfuryl isopropyl sulfide, 99%

Manufacturer: Alfa Aesar™ B2199406

Quantity 5g

 View more versions of this product

Catalog No. AAB2199406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1883-78-9
    Molecular Formula C8H12OS
    Formula Weight 156.25
    MDL Number MFCD00040265
    Beilstein 1306593
    Chemical Name or Material Furfuryl isopropyl sulfide

    Specifications

    Formula Weight 156.25
    TSCA TSCA
    Molecular Formula C8H12OS
    MDL Number MFCD00040265
    CAS 1883-78-9
    Beilstein 1306593
    Boiling Point 83° to 85°C (16mmHg)
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Quantity 5g
    Flash Point 76°C (169°F)
    Odor Garlic-like
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Chemical Name or Material Furfuryl isopropyl sulfide
    Assay 99%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit