Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Furfuryl isopropyl sulfide, 99%

Click to view available options
Quantity:
5g
25g
1883-78-9
C8H12OS
156.243
MFCD00040265
WCHRNAKORAINOJ-UHFFFAOYSA-N
furfuryl isopropyl sulfide,2-isopropylthio methyl furan,isopropyl furfuryl sulfide,furfurylisopropylsulfide,unii-5ott266rl5,fema no. 3161,furan, 2-1-methylethyl thio methyl,furan, 2-isopropylthio methyl,2-1-methylethyl thio methyl furan
61282
2-(propan-2-ylsulfanylmethyl)furan
CC(C)SCC1=CC=CO1
1883-78-9
C8H12OS
156.243
MFCD00040265
WCHRNAKORAINOJ-UHFFFAOYSA-N
furfuryl isopropyl sulfide,2-isopropylthio methyl furan,isopropyl furfuryl sulfide,furfurylisopropylsulfide,unii-5ott266rl5,fema no. 3161,furan, 2-1-methylethyl thio methyl,furan, 2-isopropylthio methyl,2-1-methylethyl thio methyl furan
61282
2-(propan-2-ylsulfanylmethyl)furan
CC(C)SCC1=CC=CO1
Flash Point 76°C (169°F)
Odor Garlic-like
Quantity 25g
Boiling Point 83°C to 85°C (16mmHg)
Beilstein 1306593
Formula Weight 156.25
Percent Purity 99%
Chemical Name or Material Furfuryl isopropyl sulfide

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA