Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfurylamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
617-89-0
C5H7NO
97.12
MFCD00003258
DDRPCXLAQZKBJP-UHFFFAOYSA-N
furfurylamine,2-furfurylamine,2-furanmethylamine,2-aminomethylfuran,2-aminomethyl furan,2-furanmethanamine,2-furylmethylamine,2-furylmethyl amine,usaf q-1,methylamine, 1-2-furyl
3438
furan-2-ylmethanamine
C1=COC(=C1)CN
617-89-0
C5H7NO
97.12
MFCD00003258
DDRPCXLAQZKBJP-UHFFFAOYSA-N
furfurylamine,2-furfurylamine,2-furanmethylamine,2-aminomethylfuran,2-aminomethyl furan,2-furanmethanamine,2-furylmethylamine,2-furylmethyl amine,usaf q-1,methylamine, 1-2-furyl
3438
furan-2-ylmethanamine
C1=COC(=C1)CN
Density 1.0900g/mL
Melting Point -70°C
Boiling Point 145°C to 146°C (761.0mmHg)
Refractive Index 1.4890 to 1.491
Flash Point 37°C
Specific Gravity 1.09
Viscosity 1.6 mPa.s (20°C)
Color Brown-Yellow to Colorless
Packaging Glass bottle
Quantity 1L
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 18, 584
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 97.12
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material Furfurylamine, 99+%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-536-9

RTECSNumber : LU9275000

TSCA : TSCA