Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (Furfurylthio)acetone, 96%

Click to view available options
Quantity:
5g
25g
58066-86-7
C8H10O2S
170.226
MFCD02683098
WWWHLSMCOBIZNV-UHFFFAOYSA-N
furfurylthio acetone,1-furan-2-ylmethylsulfanyl propan-2-one,1-furfurylthio-2-propanone,1-furan-2-ylmethyl sulfanyl propan-2-one,1-2-furylmethylsulfanyl propan-2-one,2-propanone,1-2-furanylmethyl thio,2-propanone, 1-2-furanylmethyl thio,1-2-furylmethyl sulfanyl acetone,furfurythioacetone,furfurylthioacetone
226249
1-(furan-2-ylmethylsulfanyl)propan-2-one
CC(=O)CSCC1=CC=CO1
58066-86-7
C8H10O2S
170.226
MFCD02683098
WWWHLSMCOBIZNV-UHFFFAOYSA-N
furfurylthio acetone,1-furan-2-ylmethylsulfanyl propan-2-one,1-furfurylthio-2-propanone,1-furan-2-ylmethyl sulfanyl propan-2-one,1-2-furylmethylsulfanyl propan-2-one,2-propanone,1-2-furanylmethyl thio,2-propanone, 1-2-furanylmethyl thio,1-2-furylmethyl sulfanyl acetone,furfurythioacetone,furfurylthioacetone
226249
1-(furan-2-ylmethylsulfanyl)propan-2-one
CC(=O)CSCC1=CC=CO1
Quantity 5g
Formula Weight 170.23
Percent Purity 96%
Chemical Name or Material (Furfurylthio)acetone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning