Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furosemide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
54-31-9
C12H11ClN2O5S
330.74
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
NS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl
54-31-9
C12H11ClN2O5S
330.74
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
NS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl

F1133, 54-31-9, C12H11ClN2O5S

Spectrum™ Furosemide, USP is used to reduce extra fluid in the body (edema) caused by conditions such as heart failure, liver disease, and kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.01
Quantity 25 g
CAS Max % 1
Percent Purity 98 to 102%