Learn More
MilliporeSigma™ Supelco™ Fused Silica-Lined Stainless Steel Tubing
Combines the strength of stainless steel with the inertness of deactivated fused silica
$122.00 - $912.00
Specifications
For Use With (Application) Gas Transfer Lines
Type Tubing
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Overall Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Overall Length (English) Price Quantity & Availability  
11-101-0390
View Documents
MilliporeSigma™ Supelco™
24965
0.085 in. 1/8 in. 6 ft.
Each for $122.00
Add to cart
11-100-8034
View Documents
MilliporeSigma™ Supelco™
24952
0.01 in. 1/16 in. 50 ft.
Each for $593.00
Add to cart
11-100-6868
View Documents
MilliporeSigma™ Supelco™
24953
0.01 in. 1/16 in. 100 ft.
Each for $912.00
Add to cart
11-100-9125
View Documents
MilliporeSigma™ Supelco™
24955
0.02 in. 1/16 in. 25 ft.
Each for $302.00
Add to cart
11-100-9167
View Documents
MilliporeSigma™ Supelco™
24959
0.03 in. 1/16 in. 25 ft.
Each for $292.00
Add to cart
11-100-6869
View Documents
MilliporeSigma™ Supelco™
24961
0.03 in. 1/16 in. 100 ft.
Each for $912.00
Add to cart
Description
Description
  • For use with gas transfer lines
Specifications
Specifications
Gas Transfer Lines
Tubing