Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUSIP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11093598SS

 View more versions of this product

Catalog No. NB11093598T


Add to cart

Description

Description

FUSIP1 Polyclonal antibody specifically detects FUSIP1 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

FUSIP1
1 mg/mL
Western Blot 1:1000 -1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunohistochemistry-Paraffin
O75494
40 kDa SR-repressor protein, FLJ43846, FUS interacting protein (serine-arginine rich) 1, FUS-interacting protein (serine-arginine rich) 2, FUS-interacting serine-arginine-rich protein 1, FUSIP1DKFZp686H0644, FUSIP2, neural-salient SR protein, NSSR, serine/arginine-rich splicing factor 10, serine-arginine repressor protein (40 kDa), SFRS13, SFRS13A, Splicing factor SRp38, splicing factor, arginine/serine-rich 13, Splicing factor, arginine/serine-rich 13ATLS-associated protein with Ser-Arg repeats, SR splicing factor 10, SRp38, SRrp40TLS-associated protein with SR repeats, TASR1TLS-associated SR protein, TASR2FUS interacting protein (serine/arginine-rich) 1, TASRFLJ30749, TLS-associated protein TASR, TLS-associated serine-arginine protein, TLS-associated serine-arginine protein 1, TLS-associated serine-arginine protein 2
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
10772
Human, Mouse
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
Affinity Purified
SRSF10
Synthetic peptide made to an internal portion of human FUSIP1 (within residues 100-200). [Swiss-Prot# O75494]
22 kDa
0.025 ml
Store at 4C. Do not freeze.
Polyclonal
This is specific for isoform 3 of the FUSIP1 protein.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only