Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUSIP1 Antibody
Rabbit Polyclonal Antibody
Specifications
Antigen | FUSIP1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000 -1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11093598
|
Novus Biologicals
NB11093598 |
0.1 mL |
Each of 1 for $482.50
|
N/A | |||||
Description
FUSIP1 Polyclonal specifically detects FUSIP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FUSIP1 | |
Western Blot 1:1000 -1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:2000, Immunohistochemistry-Paraffin | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
O75494 | |
10772 | |
Synthetic peptide made to an internal portion of human FUSIP1 (within residues 100-200). [Swiss-Prot# O75494] | |
Primary | |
Store at 4C. Do not freeze. | |
22 kDa |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
40 kDa SR-repressor protein, FLJ43846, FUS interacting protein (serine-arginine rich) 1, FUS-interacting protein (serine-arginine rich) 2, FUS-interacting serine-arginine-rich protein 1, FUSIP1DKFZp686H0644, FUSIP2, neural-salient SR protein, NSSR, serine/arginine-rich splicing factor 10, serine-arginine repressor protein (40 kDa), SFRS13, SFRS13A, Splicing factor SRp38, splicing factor, arginine/serine-rich 13, Splicing factor, arginine/serine-rich 13ATLS-associated protein with Ser-Arg repeats, SR splicing factor 10, SRp38, SRrp40TLS-associated protein with SR repeats, TASR1TLS-associated SR protein, TASR2FUS interacting protein (serine/arginine-rich) 1, TASRFLJ30749, TLS-associated protein TASR, TLS-associated serine-arginine protein, TLS-associated serine-arginine protein 1, TLS-associated serine-arginine protein 2 | |
SRSF10 | |
IgG | |
Affinity Purified | |
This is specific for isoform 3 of the FUSIP1 protein. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title