Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKOR2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548319

Catalog No. PA548319

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Exhibits transcriptional repressor activity. Acts as a TGF-beta antagonist in the nervous system.
Specifications

Specifications

SKOR2
Polyclonal
PBS with 0.09% sodium azide
Q2VWA4
SKI family transcriptional corepressor 2, Functional Smad-suppressing element on chromosome 18, Fussel-18, LBX1 corepressor 1-like protein, Ladybird homeobox corepressor 1-like protein, SKOR2, CORL2, FUSSEL18
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
652991
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SKOR2
Liquid
SKOR2
KLH conjugated synthetic peptide between 870-898 amino acids from the C-terminal region of human FUSSEL18.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit