Learn More
FUT10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 186601AP
Catalog No. 50-173-2189
Add to cart
Description
Description
Probable fucosyltransferase.
Specifications
Specifications
FUT10
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5F2L1, Q5F2L2, Q6P4F1
Fuc TX, Fucosyltransferase 10, Fucosyltransferase X, FucT X, FUT10
Rabbit
IgG
150 μL
-20°C
Primary
171167, 497619, 84750
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FUT10
Liquid
Fut10
FUT10 Fusion Protein Ag13415
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat