Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4 Mouse anti-Human, Clone: HI98, Abnova™

Mouse monoclonal antibody raised against human FUT4.

Manufacturer:  Abnova Corporation MAB12956

Catalog No. 89-934-174


Add to cart

Description

Description

The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq]

Specifications

Specifications

FUT4
HI98
Mouse monoclonal antibody raised against human FUT4.
In PBS, pH 7.2 (0.09% sodium azide).
CD15/ELFT/FCT3A/FUC-TIV/FUTIV
Mouse
IgM
50 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Liquid
FUT4
Human peripheral blood mononuclear cell (PBMC).
Affinity purification
RUO
Primary
2526
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit