Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4 Mouse anti-Human, Mouse, Clone: MCS-1 (PerCP), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13814

Catalog No. 89-033-879


Add to cart

Description

Description

Mouse monoclonal antibody raised against human FUT4.

  • High specificity
  • High reactivity
Specifications

Specifications

FUT4
MCS-1
Mouse monoclonal antibody raised against human FUT4.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
FUT4
X-hapten on lacto-N-fucose pentaosyl lll.
Protein A/G purification
RUO
Primary
2526
Flow Cytometry
PerCP
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Q2VLL5
CD15|ELFT|FCT3A|FUC-TIV|FUTIV
Mouse
IgG3
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)