Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548749

Catalog No. PA548749

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May catalyze alpha-1,3 glycosidic linkages involved in the expression of sialyl Lewis X antigens.
Specifications

Specifications

FUT7
Polyclonal
PBS with 0.09% sodium azide
Q11130
Alpha-(1, 3)-fucosyltransferase 7, 241-, Fucosyltransferase 7, Fucosyltransferase VII, Fuc-TVII, FucT-VII, Galactoside 3-L-fucosyltransferase, Selectin ligand synthase, FUT7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2529
Western Blot
Unconjugated
FUT7
Liquid
FUT7
KLH conjugated synthetic peptide between 78-106 amino acids from the N-terminal region of human FUT7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit