Learn More
Moore Medical Futuro™ Deluxe Wrist Stabilizer
Comfort, protection and support for injured, weak or post-cast wrists
Manufacturer:  Moore Medical 77623
 View more versions of this product
Catalog No. 19-898-600
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For use with wrist injuries, including Carpal Tunnel Syndrome, Sprains, Tendonitis, Post Cast issues, etc.
  • Slide-guard fasteners
  • Three-strap design for rigid stability
  • Palmar and dorsal (top of hand) stabilizers for additional support
  • Allows free movement of fingers
  • Palm padding uses comfortable, conforming memory foam
  • Comfort pad strap prevents irritation around thumb
  • Breathable and latex-free materials are durable and soft
  • Easy sleeve design is easy to put on and take off
Some individuals may be sensitive to neoprene or neoprene-blend rubber. If a rash develops, discontinue use and consult a physician.
Specifications
Specifications
Futuro™ Deluxe Wrist Stabilizer
Slide guard™
Provides comfortable protection and firm support to injured, weak or post-cast wrists requiring rigid stabilization like Carpal tunnel syndrome, post-cast, arthritis, sprains, tendonitis
Palm padding, Comfort Pad™ strap
5.5 to 7.5 in.
Small/Medium
Beige
Left
Neoprene
13.97 to 19.05 cm