Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical Futuro™ Deluxe Wrist Stabilizer

Comfort, protection and support for injured, weak or post-cast wrists

Specifications

Closure Type Slide guard™
Color Beige
Product Type Futuro™ Deluxe Wrist Stabilizer
Size Small/Medium
For Use With (Application) Provides comfortable protection and firm support to injured, weak or post-cast wrists requiring rigid stabilization like Carpal tunnel syndrome, post-cast, arthritis, sprains, tendonitis
View More Specs

Products 1
Catalog Number Mfr. No. Hand Price Quantity & Availability  
Catalog Number Mfr. No. Hand Price Quantity & Availability  
19-898-600
View Documents
Moore Medical
77623
Left This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

For use with wrist injuries, including Carpal Tunnel Syndrome, Sprains, Tendonitis, Post Cast issues, etc.

  • Slide-guard fasteners
  • Three-strap design for rigid stability
  • Palmar and dorsal (top of hand) stabilizers for additional support
  • Allows free movement of fingers
  • Palm padding uses comfortable, conforming memory foam
  • Comfort pad strap prevents irritation around thumb
  • Breathable and latex-free materials are durable and soft
  • Easy sleeve design is easy to put on and take off

Some individuals may be sensitive to neoprene or neoprene-blend rubber. If a rash develops, discontinue use and consult a physician.

Specifications

Specifications

Slide guard™
Futuro™ Deluxe Wrist Stabilizer
Provides comfortable protection and firm support to injured, weak or post-cast wrists requiring rigid stabilization like Carpal tunnel syndrome, post-cast, arthritis, sprains, tendonitis
Neoprene
13.97 to 19.05 cm
Beige
Small/Medium
Palm padding, Comfort Pad™ strap
5.5 to 7.5 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit