Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR/NR1H4 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4F12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271526JF549

Catalog No. N271526J549


Add to cart

Description

Description

FXR/NR1H4 Monoclonal antibody specifically detects FXR/NR1H4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

FXR/NR1H4
OTI4F12
Immunohistochemistry
Affinity Purified, Conjugated
NR1H4
Recombinant protein fragment corresponding to amino acids 21-297 of human FXR/NR1H4 (NP_005114) produced in E.coli.
Immunogen affinity purified
Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction
Primary
9971.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit