Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR Mouse anti-Human, Clone: OTI4F12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527121

Catalog No. PIMA527121


Add to cart

Description

Description

The steroid receptor superfamily acts through direct association with DNA sequences known as hormone response elements (HREs) and bind DNA as either homo- or heterodimers. The promiscuous mediator of heterodimerization, RXR, is the receptor for 9-cis retinoic acid, and dimerizes with VDR, TR, PPAR, as well as several novel receptors including LXR (also referred to as RLD-1) and FXR. FXR and LXR fall into a category of proteins termed orphan receptors because of their lack of a defined function, and in the case of LXR, the lack of a defined ligand. FXR has been shown to bind a class of lipid molecules called farnesoids. LXR/RXR heterodimers have highest affinity for DR-4 DNA elements while FXR/RXR heterodimers bind IR-1 elements. Both LXR/RXR and FXR/RXR heterodimers retain their responsiveness to 9-cis retinoic acid.
Specifications

Specifications

FXR
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96RI1
BAR, FXR, HRR-1, HRR1, RIP14, RXR-interacting protein 14, bile acid receptor, farnesoid X nuclear receptor, farnesoid X-activated receptor, farnesol receptor HRR-1, retinoid X receptor-interacting protein 14
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9971
Immunohistochemistry (Paraffin), Western Blot
OTI4F12
Unconjugated
NR1H4
Liquid
NR1H4
Human recombit protein fragment corresponding to amino acids 21-297 of NR1H4 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit