Learn More
FXR1 Mouse anti-Human, Mouse, Rat, Clone: 1E12E8, ProteinTech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 678131IG150UL

Description
Aliquoting is unnecessary for -20°C storage.
FXR1 Monoclonal antibody specifically detects FXR1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
FXR1 | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P51114, Q5XI81, Q61584 | |
Fxr1 | |
FXR1 Fusion Protein Ag27800 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1E12E8 | |
Unconjugated | |
Fxr1 | |
1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene 1, autosomal homolog; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p | |
Mouse | |
Protein G | |
RUO | |
14359, 361927, 8087 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.