Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR1 Rabbit anti-Human, Mouse, Rat, Rhesus Monkey, Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Manufacturer:  Invitrogen 711442

Catalog No. PI711442


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Bovine and Sheep Recombit rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombit monoclonal antibodies, providing the best of both worlds – the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombit antibody. While functionally the same as a polyclonal antibody – recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets – a recombit rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predomitly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

FXR1
Recombinant Polyclonal
Unconjugated
FXR1
Liquid
FXR1
Protein corresponding to Human FXR1 (aa 400-600)
Protein A
RUO
Antibody
Oligoclonal
Human, Mouse, Rat, Rhesus Monkey
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P51114, Q61584, Q5XI81
FXR1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14359, 361927, 710170, 8087
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only