Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FXYD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109744
Description
Immunogen sequence: MASLGHILVF CVGLLTMAKA ESPKEHDPFT YDYQSLQIGG LVIAGILFIL GILIVLSRRC RCKFNQQQRT GEPDEEEGTF RSSIRRLSTR RR.
This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na,K-ATPase. Alternatively spliced transcript variants have been described.
Specifications
FXYD1 | |
Polyclonal | |
Unconjugated | |
FXYD1 | |
0610012C17Rik; 1110006M24Rik; FXYD domain containing ion transport regulator 1; FXYD domain-containing ion transport regulator 1; FXYD1; Phospholemman; PLM; Pml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56188, 58971 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.46 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O08589, Q9Z239 | |
FXYD1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-92 of human FXYD1 (NP_005022.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction