Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD3 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271219AF405

Catalog No. N271219A405


Add to cart

Description

Description

FXYD3 Monoclonal antibody specifically detects FXYD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

FXYD3
OTI4E3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
FXYD3
Full length recombinant protein of human FXYD3 (NP_005962) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Chloride conductance inducer protein Mat-8, FXYD domain containing ion transport regulator 3, FXYD domain-containing ion transport regulator 3, Mammary tumor 8 kDa protein, MAT8MAT-8, Phospholemman-like, phospholemman-like protein, PLMLMGC111076
Mouse
IgG2a
0.1 mL
Primary
5349.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit