Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549345

Catalog No. PA549345

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the FXYD family of transmembrane proteins. This particular protein encodes phosphohippolin, which likely affects the activity of Na,K-ATPase. Multiple alternatively spliced transcript variants encoding the same protein have been described. Related pseudogenes have been identified on chromosomes 10 and X. Read-through transcripts have been observed between this locus and the downstream sodium/potassium-transporting ATPase subunit gamma (FXYD2, GeneID 486) locus.[provided by RefSeq, Feb 2011]
Specifications

Specifications

FXYD6
Polyclonal
PBS with 0.09% sodium azide
Q9H0Q3, Q9D164
FXYD domain-containing ion transport regulator 6, Phosphohippolin, FXYD6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
53826, 59095
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FXYD6
Liquid
FXYD6
KLH conjugated synthetic peptide between 81-115amino acids from the C-terminal region of human FXYD6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit