Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $436.00
Specifications
Antigen | FXYD6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FXYD6 Polyclonal specifically detects FXYD6 in Mouse, Rat samples. It is validated for Western Blot.Specifications
FXYD6 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
53826 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
FXYD domain containing ion transport regulator 6, FXYD domain-containing ion transport regulator 6, phosphohippolin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human FXYD6 (NP_071286.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title