Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FYCO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545805
Description
Peptide sequence: PLAQELEATR DSLDKKNQHL ASFPGWLAMA QQKADTASDT KGRQEPIPSD Sequence homology: Horse: 86%; Human: 100%.
This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).
Specifications
FYCO1 | |
Polyclonal | |
Unconjugated | |
FYCO1 | |
2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79443 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BQS8 | |
FYCO1 | |
Synthetic peptide directed towards the N-terminal of human FYCO1 (aa 346-395). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction