Learn More
FZD1 Polyclonal Antibody, Invitrogen™
Description
PA1-32766 detects Frizzled 1 from human samples. PA1-32766 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32766 immunogen is: Synthetic peptide, KLH conjugated.
Specifications
Specifications
| Antigen | FZD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Gene | FZD1 |
| Gene Accession No. | Q9UP38 |
| Gene Alias | DKFZp564G072, FLJ95923 |
| Gene Symbols | FZD1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.