Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZD7 Monoclonal Antibody (151143), Invitrogen™

Rat Monoclonal Antibody

Supplier:  Thermo Scientific MA524378

Catalog No. PIMA524378


Description

Description

In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -3, -4 or -8 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

FZD7 Monoclonal antibody specifically detects FZD7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FZD7
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O75084, Q61090
FZD7
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185
50 μg
Primary
Human, Mouse
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry
151143
Unconjugated
FZD7
frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7
Rat
Protein A/G
RUO
14369, 8324
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.