Learn More
Invitrogen™ FZD8 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547071
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Frizzled-9 is observed and less than 1% cross-reactivity with rmFrizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-4, rmFrizzled-6, and rmFrizzled-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Members of the Frizzled family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. Frizzled 8 is highly expressed in two human cancer cell lines, indicating that it may play a role in several types of cancer. Its expression has been reported in brain, heart, kidney, pancreas, and skeletal muscle. ESTs have been isolated from embryo, eye, head/neck, heart, kidney, liver/spleen, pancreas, prostate, and testis libraries, as well as many tumor libraries. The crystal structure of the extracellular cysteine-rich domain of a similar mouse protein has been determined.
Specifications
FZD8 | |
Polyclonal | |
Unconjugated | |
Fzd8 | |
frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14370 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q61091 | |
Fzd8 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-8 Ala25-Pro173. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.