Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FzM1.8

Click to view available options
Quantity:
10 mg
50 mg
Description
FzM1.8 is a frizzled 4 allosteric agonist (pEC50 = 6.4). Potentiates the β-catenin pathway in the absence of any WNT ligand, by biasing FZD4 signaling toward the FZD4/PI3K axis. Preserves stemness and promotes proliferation of undifferentiated cells.
Specifications
Specifications
Chemical Name or Material | 3-Hydroxy-5-[[(2-Naphthalenylamino)carbonyl]amino]benzoic acid |
CAS | 2204290-85-5 |
Quantity | 10 mg |
Molecular Formula | C18H14N2O4 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction