Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FZR1/CDH1 Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: AR38.2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP219018AF594

Catalog No. NP219018A94


Add to cart

Description

Description

FZR1/CDH1 Monoclonal antibody specifically detects FZR1/CDH1 in Human,Mouse samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

FZR1/CDH1
AR38.2
Western Blot, ELISA
CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FZR1
Recombinant Fizzy Related Protein
0.1 mL
Primary
51343
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit