Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ G 28UCM
FASN inhibitor
Supplier: Tocris Bioscience™ 5446/50
This item is not returnable.
View return policy
Description
G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.Specifications
3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate | |
50 mg | |
C24H16O10 | |
>98% |
1094451-90-7 | |
Fatty Acid Synthase Inhibitors | |
464.38 |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction