Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-CSF Antibody (243), Alexa Fluor™ 405
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289191AF405
Description
G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Sandwich ELISA Detection.Specifications
G-CSF | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
243 | |
Sandwich ELISA Detection | |
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human G-CSF (NP_757373.1; Ala30-Pro204). | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
1440 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction