Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Rabbit anti-Human, Macaque, Clone: CSF3/3166R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen G-CSF
Clone CSF3/3166R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123004
View Documents
Novus Biologicals
NBP30776520UG
20 μg
Each for $159.82
Add to cart
 
NB123003
View Documents
Novus Biologicals
NBP30776510UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

G-CSF Monoclonal antibody specifically detects G-CSF in Human, Macaque samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

G-CSF
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human, Macaque
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Recombinant full-length human G-CSF protein
Monoclonal
Protein A or G purified
CSF3/3166R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research
10 mM PBS with 0.05% BSA
1440
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit