Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Rat anti-Mouse, eFluor® 660, Clone: 9B4CSF, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 50735380

 View more versions of this product

Catalog No. 50-112-4454


Add to cart

Description

Description

Description: This 9B4CSF monoclonal antibody reacts with mouse G-CSF (Granulocyte Colony-Stimulating Factor), a growth factor that stimulates the proliferation, differentiation, and mobilization of hematopoetic stem cells. It is expressed by monocytes, macrophages, and bone marrow stromal cells, and can also be induced in fibroblasts by IL-17A. Unlike GM-CSF and IL-3, which can stimulate cells of multiple lineages, G-CSF activity is limited to neutrophilic granulocytes. G-CSF is essential to the maintece of neutrophil counts during homeostasis, and low basal levels of the protein are detectable in the serum of healthy individuals. Circulating levels become elevated rapidly upon infection, as G-CSF is also important for the activation and mobilization of mature neutrophils during the innate immune response. G-CSF therapy is commonly used to treat neutropenia following bone marrow transplant or chemotherapy. This 9B4CSF antibody has been demonstrated to cross-react with human G-CSF. Applications Reported: This 9B4CSF antibody has been reported for use in flow cytometric analysis, and intracellular staining followed by flow cytometric analysis. Applications Tested: This 9B2-8°CSF antibody has been tested by intracellular staining followed by flow cytometric analysis of in vitro-cultured and stimulated mouse monocytes/macrophages. This can be used at less than or equal to 0.06 µg per test.

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet ft.s syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
Specifications

Specifications

G-CSF
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P09920
C17orf33, CSF3OS, G-CSF, GCSF, MGC45931
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
9B4CSF
eFluor™ 660
Csf3
Liquid
Csf3
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12985
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.