Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G gamma3 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen G gamma3
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094758
View Documents
Novus Biologicals
NBP303515100UL
100 μg Each for $320.71
Add to cart
 
NB094759
View Documents
Novus Biologicals
NBP30351520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

G gamma3 Polyclonal antibody specifically detects G gamma3 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

G gamma3
Unconjugated
Rabbit
Mouse, Rat
GNGT3, guanine nucleotide binding protein (G protein), gamma 3, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3, guanine nucleotide-binding protein gamma-3 subunit, NBP gamma-3
Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human G gamma3 (NP_036334.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
2785
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit