Learn More
Novus Biologicals g-Linked ATP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
Manufacturer:  Novus Biologicals 5020005
 View more versions of this product
Catalog No. 50-200-05K
  Check Availability
Add to cart
Specifications
Description
Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.
Recommended Storage
Store at 4°C. Do not freeze.
Description & Specifications
Specifications
Low ligand substitution (1-2μmol/mL)
g-Linked ATP Agarose Affinity Resin
5mL