Novus Biologicals g-Linked ATP Agarose Affinity Resin

Affinity resin for purification of enzymes that bind nucleotides and related molecules

Manufacturer: Novus Biologicals 5100005

 View all options for this product

Catalog No. 51-000-05

  Check Availability
Add to Cart


    Formulation High ligand substitution (8-12μmol/mL)
    Product Type g-Linked ATP Agarose Affinity Resin
    Quantity 5mL

    Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.

    • ATP Agarose resin comprises ATP attached to agarose beads via its gamma-phosphate
    • Two forms of resin are available with low and high ligand substitution
    • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules

    Recommended Storage

    Store at 4°C. Do not freeze.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit