Novus Biologicals g-Linked GTP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
Manufacturer: Novus Biologicals 5050002
 View more versions of this product
Catalog No. 50-500-02K
  Check Availability
Add to Cart
    Specifications
    Product Type g-Linked GTP Agarose Affinity Resin
    Quantity 5mL
    • GTP Agarose resin comprises GTP attached to agarose beads via its gamma-phosphate
    • Resin is resistant to phosphatases found in many crude tissue extracts
    • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules
    Recommended Storage
    Store at 4°C. Do not freeze.
  • Description & Specifications
Related Products
Product Content Feedback