Novus Biologicals g-Linked GTP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
Manufacturer: Novus Biologicals 5050001
 View more versions of this product
Catalog No. 50-500-01K
  Check Availability
Add to Cart
    Specifications
    Product Type g-Linked GTP Agarose Affinity Resin
    Quantity 2mL
    Recommended Storage
    Store at 4°C. Do not freeze.
  • Description & Specifications
Related Products
Product Content Feedback