Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein alpha 12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | G protein alpha 12 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB15743020
![]() |
Novus Biologicals
NBP15532120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155321
![]() |
Novus Biologicals
NBP155321 |
100 μL |
Each for $487.50
|
|
|||||
Description
G protein alpha 12 Polyclonal specifically detects G protein alpha 12 in Human samples. It is validated for Western Blot.Specifications
G protein alpha 12 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
G alpha-12, gep, G-protein subunit alpha-12, guanine nucleotide binding protein (G protein) alpha 12, guanine nucleotide-binding protein subunit alpha-12, MGC104623, MGC99644, NNX3, RMP, WUGSC:H_GS165O14.2 | |
GNA12 | |
IgG | |
This product is specific to Subunit or Isoform: alpha-12. |
Western Blot | |
Unconjugated | |
RUO | |
Q03113 | |
2768 | |
Synthetic peptides corresponding to GNA12(guanine nucleotide binding protein (G protein) alpha 12) The peptide sequence was selected from the middle region of GNA12. Peptide sequence TFQLYVPALSALWRDSGIREAFSRRSEFQLGESVKYFLDNLDRIGQLNYF. | |
Primary | |
44 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title