Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha 12 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP303329100UL

 View more versions of this product

Catalog No. NB092805


Add to Cart

Description

Description

G protein alpha 12 Polyclonal specifically detects G protein alpha 12 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

G protein alpha 12
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat
Purified
Western Blot, Immunofluorescence
Unconjugated
G alpha-12, gep, G-protein subunit alpha-12, guanine nucleotide binding protein (G protein) alpha 12, guanine nucleotide-binding protein subunit alpha-12, MGC104623, MGC99644, NNX3, RMP, WUGSC:H_GS165O14.2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human G protein alpha 12 (NP_031379.2).
100 μL
Signal Transduction
2768
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit