Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha 16 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI6B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270836AF647

Catalog No. N270836A647


Add to cart

Description

Description

G protein alpha 16 Monoclonal antibody specifically detects G protein alpha 16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

G protein alpha 16
OTI6B3
Western Blot
Affinity Purified, Conjugated
GNA15
Recombinant protein fragment corresponding to amino acids 1-256 of human G protein alpha 16 (NP_002059) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
2769.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
G alpha-15, G alpha-16, GNA16G-protein subunit alpha-15, G-protein subunit alpha-16, guanine nucleotide binding protein (G protein), alpha 15 (Gq class), guanine nucleotide-binding protein subunit alpha-15, Guanine nucleotide-binding protein subunit alpha-16
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit