Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI12D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270837AF700

Catalog No. N270837A700


Add to cart

Description

Description

G protein alpha Monoclonal antibody specifically detects G protein alpha in Human samples. It is validated for Western Blot.
Specifications

Specifications

G protein alpha
OTI12D9
Western Blot
Affinity Purified, Conjugated
GNAS
Full length recombinant protein of human G protein alpha (NP_000507) produced in HEK293T cells.
Immunogen affinity purified
Signal Transduction
Primary
2778.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Adenylate cyclase-stimulating G alpha protein, AHO, Alternative gene product encoded by XL-exon, Extra large alphas protein, GNAS complex locus, GNAS1GSA, GNASXL, GPSAC20orf45, GSP, guanine nucleotide binding protein (G protein), alpha stimulating activitypolypeptide 1, guanine nucleotide regulatory protein, guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas, MGC33735, NESP, NESP55, neuroendocrine secretory protein, PHP1A, PHP1B, PHP1C, POH, protein ALEX, SCG6, secretogranin VI, XLalphas
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit