Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein beta 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10098706
Description
G protein beta 4 Polyclonal specifically detects G protein beta 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray.Specifications
G protein beta 4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from whole antisera with No Preservative | |
G protein beta-4 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 4, guanine nucleotide binding protein beta subunit 4, guanine nucleotide-binding protein subunit beta-4, Transducin beta chain 4 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
GBB4. | |
Human, Mouse, Rat | |
Antisera |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
LYOPH | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Microarray | |
Q9HAV0 | |
GNB4 | |
A synthetic peptide from internal region of human G protein beta 4 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat, mouse and Xenopus species. | |
0.1 mL | |
Signal Transduction | |
59345 | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction