Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein beta 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$176.85 - $436.00
Specifications
Antigen | G protein beta 4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1: 300 - 1: 2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1: 300 - 1: 2000 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10098706
|
Novus Biologicals
NB10098706 |
0.1 mL |
Each for $436.00
|
|
NB017942
|
Novus Biologicals
NB100987060.025ML |
0.025 mL |
Each for $176.85
|
|
Description
G protein beta 4 Polyclonal specifically detects G protein beta 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray.Specifications
G protein beta 4 | |
Western Blot 1: 300 - 1: 2000, Immunohistochemistry 1: 300 - 1: 2000, Immunohistochemistry-Paraffin 1: 300 - 1: 2000 | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
Whole antisera with No Preservative | |
G protein beta-4 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 4, guanine nucleotide binding protein beta subunit 4, guanine nucleotide-binding protein subunit beta-4, Transducin beta chain 4 | |
GNB4 | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
Q9HAV0 | |
59345 | |
A synthetic peptide from an internal part of human GNB4 conjugated to an immunogenic carrier protein was used as the immunogen. The peptide is homologous in rat, mouse and xenopus. | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
GBB4. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title