Learn More
MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor XXII , Aβ42-Selective, CS-1
The γ-Secretase Inhibitor XXII, Aβ42-Selective, CS-1, also referenced under CAS 1210283-33-2, controls the biological activity of γ-Secretase.
Manufacturer:  MilliporeSigma™ 56579110MG
Catalog No. 56-579-110MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
1210283-33-2
466.4g/mol
White solid
10mg
C25H13F3O6
DMSO