Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-substrate Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271645AF405

Catalog No. N271645A405


Add to cart

Description

Description

G-substrate Monoclonal antibody specifically detects G-substrate in Human samples. It is validated for Western Blot.
Specifications

Specifications

G-substrate
OTI2A8
Western Blot
Affinity Purified, Conjugated
PPP1R17
Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
10842.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
chromosome 7 open reading frame 16, GSBSG-substrate
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit