Learn More
BioVision IncSupplier Diversity Partner Glycerol-3-Phosphate (G3P) Colorimetric Assay Kit: Glycerol-3-Phosphate (G3P) Colorimetric Assay Kit: For measuring G3P in animal tissues as well as adherent or suspension cells. Detection limit: less than 2 nmol. 100 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K641100
Glycerol-3-phosphate (G3P) is an important intermediate for all living organisms. Glycerol-3-Phosphate is produced either by glycerol via glycerol kinase or by dihydroxyacetone phosphate through glycerol-3-phosphate dehydrogenase. In response to cellular signals, glycerol-3-phosphate can be utilized in multiple pathways: it can be further converted into glyceraldehyde-3-phosphate and enter glycolysis or rapidly generate NAD+ in brain or muscle tissues through the G3P shuttle or enter the lipid biosynthetic pathway. Recent studies have found that glycerol-3-phosphate is a novel regulator and plays a fundamental defense role in plant pathogenesis. This kit is a sensitive, fast and easy-to-use kit. In this assay, G3P is oxidized by G3P Enzyme Mix to form an intermediate, which reduces a nearly colorless probe to a colored product with strong absorbance at 450 nm. This assay kit can detect G3P less than 2 nmol/well and can be used for a variety of sample types.
Catalog No. 50-111-9811